Learn More
Bioworld Magentaâ„¢ GA-7-3 Plant Culture Box w/o Lid, 1 CS
Supplier: Bioworld 309300091
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Magenta GA-7-3 Plant Culture Box without LidMagenta GA-7 Plant Culture Boxes are manufactured from polycarbonate, to be both autoclavable and resuable Non-sterile boxes are excellent for use in the plant culture protocols Sturdy and leak free The clear surface allows for sample viewing without disrupting growth Boxes have a growing area of 6.125 sq. inches Dimensions: 3" x 3" x 3" Sold as 100/case Please note:This product's shipping cost is calculated based on the dimensions of the largest package size. All shipping charges will be adjusted internally upon receipt of the order to accurately reflect the standard carrier costs associated with the package sizes ordered.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.