Learn More
V&P Scientific Inc MAGNETIC PLATE, 24 or 96 Well, Standard or Deep Well, Round or Conical or Pyramid Bottom, 7 Bars, North/South Magnetic Orientation
Supplier: V&P Scientific Inc VP 771MWZM1ALT
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
MAGNETIC BEAD SEPARATION PLATE, 24 or 96 Well, Standard or Deep Well Plate, Round or Conical or Pyramid Bottom, 7 Bars, 50MGO, NdFeB Magnets, White Polycarbonate Magnet Frame, 1 Sided Pellet Location, Bar 3mm Wide, Axial - Alternating North-To-South Magnetic Orientation, Center Magnet with Notch, SLAS Footprint, Includes Registration Base
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.