Learn More
Cytiva Macrosep Advance Centrifugal Devices with Omega Membrane - 100K, clear (100/pkg)
Supplier: Cytiva MAP100C38
Quickly Concentrates Up To 20 mL of Biological Sample-Rapidly concentrates 20 mL sample volumes to 0.5 mL-Achieve 50X concentration and > 90% recovery in just minutes-Low protein-binding Omega™ membrane and polypropylene housing minimize losses due to non-specific binding-Versatile Omega membrane is available in a variety of MWCOs-Built-in deadstop prevents spinning to dryness-Color-coded and laser etched for easy identification This product is not approved for use in medical, clinical, surgical or other patient applications.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.