Learn More
Biocare Medical, LLC MACH 4 Universal HRP-Polymer - 100 ml
Supplier: Biocare Medical, LLC M4U534L
The Universal HRP-Polymer detects both mouse and rabbit antibodies.This biotin-free technology uses a specific probe to detect mouse primary antibodies and is then followed by a horseradish peroxidase polymer (HRP) that binds to the probe. This innovative HRP-polymerization technology provides increased staining sensitivity for mouse monoclonal antibodies when compared to Envision and other polymer detection kits. It can be used manually and on automated stainers.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.