Learn More
Thermo Scientific™ MabCaptureC™ RoboColumn
MabCaptureC RoboColumns™ are small chromatography columns used for fully automated and parallel chromatographic separations on robotic liquid handling workstations.
$546.00 - $818.00
Specifications
Diameter (Metric) | 5 mm |
---|---|
Particle Size | 75 ±10 μm |
Ligand Type | Recombinant C domain of Protein A |
Matrix | Agarose-based, Epoxide-activated |
Max. Operating Pressure | ≤3 bar |
Catalog Number | Mfr. No. | Bed Volume | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Bed Volume | Price | Quantity & Availability | |||||
59-436-62200
|
Thermo Scientific™
5943662200 |
200 μL |
Each for $546.00
|
|
|||||
59-436-62600
|
Thermo Scientific™
5943662600 |
600 μL |
Each for $818.00
|
|
|||||
Description
MabCaptureC MiniChrom columns are designed for bench-scale resin screening, process development, and sample preparation. These pre-packed columns are ready to connect and have the following features:
- Packed with MabCaptureC Affinity Matrix, offering high dynamic binding capacity and high resolution over a wide range of flow rates
- Direct connection to standard chromatography systems
The 1 mL columns can be used for quick screenings of application feasibility and lab scale purification on a convenient and easy-to-use pre-packed column. The 10-cm bed height of the 5 mL column allows initial process development at a bench scale.
The pre-packed MabCaptureC Affinity Matrix contains engineered rProtein A that binds at the CH2-CH3 interface of the Fc-region of IgGs and shows cross binding to the VH3 domain of IgGs.
Features of MabCaptureC Affinity Matrix include:
- High dynamic mAb binding capacity of > 50 g/L at 4.8 min residence time
- Purification of mAbs and a subpopulation of scFv's and Fab fragments containing VH3 binding domains
- Clean and sanitize with standard sodium hydroxide agents for chromatography products
- Excellent scalability
- Non-animal derived
Main resin characteristics:
- Matrix: agarose-based, epoxide activated
- Average particle size: 75 ± 10 μm
- Ligand: recombinant C domain of Protein A
- Ligand coupling method: epoxide coupling
- Binding capacity: >55 g IgG/L resin at 5 min residence time and 10% breakthrough; >75 g IgG/L resin at 10 min. residence time and 10% breakthrough
- Elution conditions: 0.1 M glycine pH 3.0, or 20–100 mM sodium citrate or acetate, pH 3.0
- Cleaning conditions: 0.2 M sodium hydroxide for 15 minutes every cycle
- Column dimensions: 0.8 cm D x 2 cm L (1 mL); 0.8 cm D x 10 cm L (5 mL)
- Operating pressure: ≤ 3 bar
- Formulation buffer: 20% (v/v) ethanol
Specifications
5 mm | |
Recombinant C domain of Protein A | |
≤3 bar | |
IgG mAbs, and a subpopulation of scFv and Fab containing VH3 domain |
75 ±10 μm | |
Agarose-based, Epoxide-activated | |
Agarose bead | |
8-well strip |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.