Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ MabCaptureC™ MiniChrom Column

MabCaptureC MiniChrom columns are designed for bench-scale resin screening, process development, and sample preparation.

$584.00 - $1758.00

Specifications

Binding Capacity >55 g IgG/L resin (5 min. residence time and 10% breakthrough), >75 g IgG/L resin (10 min. residence time and 10% breakthrough)
Diameter (Metric) 0.8 cm
Particle Size 75 ±10 μm
Ligand Type Engineered rProtein A
Matrix Agarose-based, Epoxide-activated
View More Specs

Products 2
Catalog Number Mfr. No. Bed Volume Price Quantity & Availability  
Catalog Number Mfr. No. Bed Volume Price Quantity & Availability  
59-436-62001
View Documents
Thermo Scientific™
5943662001
1 mL
Each for $584.00
Add to Cart
 
59-436-62005
View Documents
Thermo Scientific™
5943662005
Each for $1,758.00
Add to Cart
 
Description

Description

MabCaptureC MiniChrom columns are designed for bench-scale resin screening, process development, and sample preparation. These pre-packed columns are ready to connect and have the following features:

  • Packed with MabCaptureC Affinity Matrix, offering high dynamic binding capacity and high resolution over a wide range of flow rates
  • Direct connection to standard chromatography systems

The 1 mL columns can be used for quick screenings of application feasibility and lab scale purification on a convenient and easy-to-use pre-packed column. The 10-cm bed height of the 5 mL column allows initial process development at a bench scale.

The pre-packed MabCaptureC Affinity Matrix contains engineered rProtein A that binds at the CH2-CH3 interface of the Fc-region of IgGs and shows cross binding to the VH3 domain of IgGs.

Features of MabCaptureC Affinity Matrix include:

  • High dynamic mAb binding capacity of > 50 g/L at 4.8 min residence time
  • Purification of mAbs and a subpopulation of scFv's and Fab fragments containing VH3 binding domains
  • Clean and sanitize with standard sodium hydroxide agents for chromatography products
  • Excellent scalability
  • Non-animal derived

Main resin characteristics:

  • Matrix: agarose-based, epoxide activated
  • Average particle size: 75 ± 10 μm
  • Ligand: recombinant C domain of Protein A
  • Ligand coupling method: epoxide coupling
  • Binding capacity: >55 g IgG/L resin at 5 min residence time and 10% breakthrough; >75 g IgG/L resin at 10 min. residence time and 10% breakthrough
  • Elution conditions: 0.1 M glycine pH 3.0, or 20–100 mM sodium citrate or acetate, pH 3.0
  • Cleaning conditions: 0.2 M sodium hydroxide for 15 minutes every cycle
  • Column dimensions: 0.8 cm D x 2 cm L (1 mL); 0.8 cm D x 10 cm L (5 mL)
  • Operating pressure: ≤ 3 bar
  • Formulation buffer: 20% (v/v) ethanol
Specifications

Specifications

>55 g IgG/L resin (5 min. residence time and 10% breakthrough), >75 g IgG/L resin (10 min. residence time and 10% breakthrough)
75 ±10 μm
Agarose-based, Epoxide-activated
Agarose bead
0.8 cm
Engineered rProtein A
≤3 bar
IgG mAbs, and a subpopulation of scFv and Fab containing VH3 domain
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Provide Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit