Learn More
METTLER TOLEDO Transmitter M800 Process 4-CH
Supplier: METTLER TOLEDO 52121853
The four-channel M800 Process is a multi-parameter transmitter that provides high-performance, reliable measurement with a convenient full-color touch screen and the ability to integrate with a wide range of ISM and analog sensors