Learn More
HF Scientific Inc M100+ White Light Laboratory Turbidimeter, 100 - 240 VAC
Supplier: HF Scientific Inc 28060
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
The white light M100+ laboratory turbidimeter offers an easy solution for verifying operation of online turbidimeters. The color touchscreen interface provides fast access to settings, data and calibration functions. Features such as the Auto Alert Calibration Prompt simplify maintenance. With a resolution of up to 0.0001 NTU at low turbidity readings and an extended range up to 4,000 NTU, the M100+ is an ideal and affordable tool for research as well as routine analytical measurements.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.