Learn More
Hach Company m-Endo Agar LES, Dehydrated 500 g
Supplier: Hach Company 2406634
m-ENDO Agar LES is a medium used for enumeration of coliforms in water using the membrane filtration procedure.