Learn More
Hach Company m-ColiBlue24 Prepared Agar Plates, pk/15.
Supplier:  Hach Company 2805215
Encompass_Preferred
In 24 hours, Hach's m-ColiBlue24 Broth give you simultaneous results for total coliforms and E. coli. Hach's ready-to-use media eliminates measuring, mixing, and autoclaving steps necessary to prepare dehydrated media. Glass ampules (26084-20), plastic ampules (260845-00), 100mL bottles (26084-42) of m-ColiBlue24 and agar plates (28052-15), afford maximum shelf life and ease of use for pre-measured and prepared media. Enumerate total coliforms and E. coli on one petri dish. Differentiate colonies easily - red and blue indicate total coliforms and blue specifies E. coli. Use m-ColiBlue24 Broth to accurately monitor and evaluate total coliform and E. coli in drinking water; wastewater; bottled water; beverages; surface, ground, and well water; and ultrapure, chemical processing and pharmaceutical processing waters.m-ColiBlue24 should only be used with pads manufactured by the following companies/brands: Pall Corporation, Sartorius, and Millipore EO.
Catalog No. 50-199-6700