Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Quidel Lyra Direct SARS COV-2 Assay
A real-time RT-PCR assay intended for the in vitro qualitative detection of human coronavirus SARS-CoV-2 from viral RNA extracted from nasal, nasopharyngeal (NP) or oropharyngeal (OP) swab specimens from patients with signs and symptoms of COVID-19.
Supplier: Quidel M941
This item is not returnable.
View return policy
Description
- The Assay targets the non-structural Polyprotein (pp1ab) of the SARS-CoV-2 virus
- One-step reagent set-up
- Results in less than 70 minutes after extraction
- Simplified and uniform workflow with standard pipetting volumes
- No freezer needed. 2°C to 8°C storage
Specifications
Yes | |
SARS-CoV-2 | |
96-well Plate | |
Control Assay |
High | |
No | |
Yes | |
Sterile |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction