Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LY 333531 hydrochloride, Tocris Bioscience™
Click to view available options
Quantity:
1 mg
Description
Isozyme-selective inhibitor of protein kinase C (PKC); competitively and reversibly inhibits PKCβI and PKCβII (IC50 values are 4.7 and 5.9 nM respectively). Selective for PKCβ over other PKC isozymes (IC50 values are 0.052, 0.25, 0.30, 0.36, 0.60 and >100 μM for PKCη, -δ, -γ, -α, -ε and -ζ respectively). Exhibits selectivity for PKC over other ATP-dependent kinases, including protein kinase A, casein kinase and src).
Specifications
Quantity | 1 mg |
Formula Weight | 505.01 |
Percent Purity | >98% |
Chemical Name or Material | LY 333531 hydrochloride |
Safety and Handling
Recommended Storage | Store at -20°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction