Learn More
Yamato Scientific America, Inc. Yamato Laboratory Stirrer, Max speed of 1200rpm for low to medium viscosity samples. Complete with stand and rod, stirring shaft and 4-blade propeller. 100V-125V
Supplier: Yamato Scientific America, Inc. LT400CA
Laboratory stirrer with high speed to support different applications for low to medium viscosity samples. Highly sensitive feedback system keeps the set speed, even with changing viscosity during stir. Maintenance free DC brushless motor. Digital speed indicator for accurate speed setting and confirmation. Noise prevention measures for optimal work environment. More safety-oriented design. Set includes stand, rod, stainless steel shaft and 60mm 4-blade propeller. Speed range 25~1200 rpm.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.