Learn More
NABERTHERM INC he muffle furnaces are for daily laboratory use. They stand out for their excellent workmanship and high level of reliability. Comes equipped with a lift door.
Supplier: NABERTHERM INC L604H2ON2480
The muffle furnaces are for daily lab use, outstanding workmanship, high level of reliability, non-classified fiber insulation, housing made of structured stainless steel, heating from two sides (three sides L24/11-LT60/12), either flap (L) or lift door (LT)