Learn More
Labrepco Inc LS3000 LABSYSTEM (W/ RACKS W/
Supplier: Labrepco Inc LS3000
Storage unit, Cryogenic sample; Labrepco; With 6 racks; Does not include boxes (holds 5 x 5 in. 100 vial boxes); Capacity: 3000 x 2.0mL vials; Overall height: 28.8 in.; Overall dia.: 26.9 in.