Learn More
Electron Microscopy Sciences LR White Accelerator 10 ML
Supplier: Electron Microscopy Sciences 14385
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
For use with LR Resin, a very low viscosity (8cps), non-toxic resin, suitable for both Light and Electron Microscopy.