Learn More
Fuel Cell Earth LQ-1105 – 1100 EW at 5% weight
Supplier: Fuel Cell Earth NS0525
LQ-1105 – 1100 EW at 5% weightDissolved Nafion™ resins in a mixture of water and alcohols1100 EWTechnical Properties: 5% wt. LIQUION™Composition: Nafion™ 4.95%-5.05% wt.Water: 20% wt.Alcohol: 75% wt.Density: 0.86 grams/cm³Volumes:Available in 25 ml, 125 mllarger volumes: Please contact us for larger volumes, e.g. 250 ml, 500 ml, 1 Liter, 1 Gallon and 15 Gallons quantities.Application:Can be dried to form films and coatings of Nafion™ or mixed with other solids to form electrodes.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.