Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
STERIS™ LpH™ III st Phenolic Sterile Disinfectant
The latest generation, low pH disinfectant
Supplier: STERIS™ 1S1808
Description
- A liquid concentrate specifically formulated for the disinfection of hard, non-porous surfaces
- Use-dilution of rate of 0.8% v/v (1:128), solution can be applied manually or sprayed
- Phenolic active ingredients provide broad spectrum antimicrobial efficacy against common bacteria and fungi
- Can be used alone or in rotation with the Vesphene™ III Disinfectants
- Ideal for use in a contamination control rotation, formulation includes surfactants for surface cleaning while disinfecting
- Products include an extensive documentation package to meet validation objectives
- Supported by the STERIS Technical Services team for individualized application and validation assistance
- Each lot documented by Certificate of Analysis
Specifications
Phenolic Disinfectant | |
Bottle | |
4/Cs. | |
3.8 L |
0.8% v/v (1:128) | |
Disinfecting Hard, Non-Porous Surfaces | |
1 gal. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction