Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Labconco™ Lower Test Tube Spindle Rack Conversion Kit



For use with SteamScrubber™, FlaskScrubber™, and FlaskScrubber™ Vantage™ Series Laboratory Glassware Washers. Labconco™ Lower Test Tube Spindle Rack Conversion Kit is used to convert existing washer rack specifically for test tubes, vials, and similar glassware.
Manufacturer: Labconco™ 4546001/EMD
Description
- Hold test tubes, vials and other laboratory glassware containers up to 10mm in diameter
- Water and detergent are injected through spindles to clean and rinse inside of tubes
- When used in FlaskScrubber or FlaskScrubber Vantage Series Washer, hot air is forced through spindles to dry inside of test tubes
- Material: Type 304 stainless steel
- Includes 150 each spindles, 5cm (2.0 in.) height and 3mm (0.125 in.) OD with nylon fasteners and riser
- Riser, when placed on top of spindles, elevates tubes 19mm (0.75 in.) from bottom of spindles to allow for proper drainage
- Requires Lower Standard Rack 4588500 (not included)
Warranty and Services
One year
Specifications
Stainless Steel (304) | |
SteamScrubber, FlaskScrubber, and FlaskScrubber Vantage Series Glassware Washers | |
12.7 in. | |
4.2 in. | |
17.5 in. | |
15 lb. |
Washer Rack Conversion Kit | |
One rack, 150 spindles with nylon fasteners, and 1 riser | |
32.2 cm | |
10.6 cm | |
44.4 cm | |
6.8 kg |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title