Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytiva Lower Section
This Lower Section is a replacement part for Whatman™ GV 050 Series Vacuum Filtration Manifolds.
Supplier: Cytiva 10442002
Description
- Description: Replacement Part for GV 050 Series Glass Vacuum Filtration Manifolds
- For Use With (Application): Vacuum Filtration
Specifications
Autoclavable | |
Reusable | |
Borosilicate Glass | |
Vacuum Filtration | |
1 Ea. |
Filter Funnel (Base Section) | |
Whatman GV050/0 Glass Vacuum Filter Holder | |
Non-sterile | |
GV050/0/02 | |
Filter Funnel |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction