Learn More
Malvern Panalytical Low Vol Disp Sizing Cell Kit
Supplier: Malvern Panalytical ZSU1002
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
Low Volume Disposable Sizing Cell Kit. Allows low volume (3uL minium sample) high quality size measurements on Zetasizer Ultra & Zetasizer Lab and allows use of extended size analysis mode and general accuracy improvements to upper particle size range without sample modification. Contains non-disposable cell anodized aluminium holder with 44 pcs. Capillary cuvettes, sealing compound and forceps
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.