Learn More
Supelco Inc LOW ADSORPTION (LA) QSERTVIAL
Supplier: Supelco Inc 29661U
volume 0.3 mL, QsertVial, clear glass vial, natural PTFE/silicone septa, thread for 9 mm
This item is not returnable.
View return policy