Learn More
FENWAL INC Lovo Valve and Filter 6/CS
SDP
Supplier:  FENWAL INC X6R4917
Encompass
This item has a minimum qty of 3 per supplier requirements.
For use with Lovo version 3.0 software and product code X6R4909 0.2 m filter with female luer, check valve, and male luer with cap For reagent addition and/or sampling during cell processing Sterile fluid path Individually packaged in plastic overwraps
Catalog No. 50-196-1849