Learn More
FENWAL INC LOVO KIT WITH BAG ACCESS(A)6CS
Supplier:  FENWAL INC X6R4909A
Encompass
Compatible with Lovo software version 3.0(1) Spinning membrane separation device with 21 μm membrane thickness and 4 μm diameter pores(1) Source product line with a 0.2 μm kit check vent(2) Solution lines, each a with solid plug(1) 800 mL In-Process bag with a swabable, needleless injection/extraction site(1) 800 mL Retentate bag with a twist-off spike port and a swabable, needleless injection/extraction site(1) 2.5 L Filtrate bag with a swabable, needleless injection/extraction site and a connection line with solid plug(1) Female luer pressure pod
Catalog No. NC1892815