Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lovibond™ Type 2 Multi-Vials
Lovibond Type 2 Multi-Vials, Includes: Lids, Plastic, Sample Vial, Diameter Inner: 0.39 in., 10mm, Graduations: 5 and 10mL, Quantity: 12 Pack, Shape: Round
Supplier: Lovibond™ 197600
Description
Lovibond Type 2 Multi-Vials, Includes: Lids, Plastic, Sample Vial, Diameter Inner: 0.39 in., 10mm, Graduations: 5 and 10mL, Quantity: 12 Pack, Shape: Round
Specifications
0.39 in. | |
Plastic | |
Lids | |
12 Pk. | |
SC450 Chlorine Colorimeter |
10 mm | |
5 and 10 mL | |
Round | |
Multi-Vial |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction