Learn More
Lovibond™ Comparator Kit to Measure Gardner (Varnish) Color Scale
Hand-held and completely portable instrument ideal for use in the field, lab or plant
Supplier:  Lovibond™ 433345
Catalog No. 09-053-437
Add to Cart
Includes: Comparator 705, two color discs (620C-45 and 620C-46), two sample cells and instructions.
Description
Description
  • Incorporates a magnifying prism which brings the color standards and the test sample side-by-side into one image, making it easy for the user to make an exact color match
  • Uses 18 permanent colored glass standards in two discs ranging from values of 1.0 to 9.0 and 9.0 to 18.0
Compliance
Meets the specifications for ASTM D-1544 and AOCS Method Td 1a.
Specifications
Specifications
ASTM D-1544
Comparator 705, two color discs (620C-45 and 620C-46), two sample cells and instructions.
1 to 18
Product Suggestions
Product Suggestions