Learn More
Lonza™ IsoGel™ IEF Plates
Precast agarose IEF plates provide rapid and reproducible isoelectric focusing of proteins up to 2000kDa
Supplier: Lonza™ 56018
Description
- Specifically designed for antibody analysis of
- Each gel is supported on GelBond™ film for easy handling and dimensional stability
- Gel size is compatible with most IEF chambers
- 125 x 100mm
Isoelectric focusing, antibody separation and analysis
Order Info
Accessories packs (Cat. No. BMA56014 and BMA56010) and blotting paper (BMA56007) available separately.
Specifications
6 to 10.5 | |
Isoelectric Focusing | |
Isoelectric focusing, Antibody separation and analysis |
Gel Plate | |
Solid | |
2° to 8°C |