Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lonza™ IsoGel™ IEF Plate Accessories
Accessories for precast agarose IEF plates. Blotting paper and accessory packs.
$173.72 - $656.50
Specifications
Separation Type | Isoelectric Focusing |
---|---|
For Use With (Application) | Isoelectric focusing, Antibody separation and analysis |
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
BMA56014
|
Lonza™
56014 |
Accessory Pack |
Each for $173.72
|
|
|||||
BMA56007
|
Lonza™
56007 |
Blotting paper |
Each for $656.50
|
|
|||||
BMA56010
|
Lonza™
56010 |
Electrophoresis Accessory Bulk Pack |
Each for $585.80
|
|
|||||
Description
- Blotting paper is for use with precast agarose IEF plates
- Accessory pack includes masks, 100mm and 125mm wicks and blotting paper
- Accessory bulk pack includes 125mm wicks and blotting paper
Specifications
Isoelectric Focusing | |
Isoelectric focusing, Antibody separation and analysis |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title