Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lonza™ BioWhittaker™ McCoy's 5A Medium (Modified)

General purpose medium for primary and established cell lines. BioWhittaker™ McCoy's 5A Medium (Modified) is manufactured in accordance with cGMP regulations.

Specifications

Product Type McCoy's 5A
Sterility Sterile
Sterilization Method Sterile-filtered through two consecutive 0.2 μm filters
Quantity 500 mL
View More Specs

Products 2
Catalog Number Mfr. No. pH Price Quantity & Availability  
Catalog Number Mfr. No. pH Price Quantity & Availability  
BW12-168F
View Documents
Lonza™
12168F
7.03 to 7.51 Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
BW12-688F
View Documents
Lonza™
12688F
6.89 to 7.2 Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
Description

Description

  • Formulated for the nutritional requirements of Walker 256 carcinoma
  • Unique components include asparagine, 3X normal glucose, amino acids (all L-isomers) and increased folic acid
  • Manufactured in accordance with cGMP regulations
  • Formulations used for standard liquid media products are those recommended by Tissue Culture Association
Specifications

Specifications

McCoy's 5A
Sterile
Sterile-filtered through two consecutive 0.2 μm filters
500 mL
SDS
Documents

Documents

Product Certifications
Provide Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit