Learn More
Medchemexpress LLC Loncastuximab, 10mg (Synonyms: RB4v1.2)
Supplier: Medchemexpress LLC HYP9971110MG
Loncastuximab (RB4v1.2) is an anti-CD19 monoclonal antibody. Loncastuximab shows antitumor activity and has potential application in non-Hodgkin's lymphoma (NHL), including diffuse large B-cell lymphoma (DLBCL)[1][2].