Learn More
LogIT Microsense Voltage and Current Sensors
SCI_ED
Especially designed to work with the DataMeter, LIVE, and LogIT family of products.
Supplier:  DCP Microdevelopment Limited CRD130545W
Catalog No. S20132ND
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Identify themselves to datalogger as soon as they are plugged in
  • Automatically calibrate and make the most effective use of the analog or digital information
  • Most are self-contained within a metal housing that encloses and protects the circuitry inside
  • Remote leads available for more flexible experiment setup
  • Designed specifically for use in schools
  • Strong Din-type connections that have proven their reliability over many years
Voltage Measurement set measures general voltage in electronic circuits or for connecting to other sensors. Resolution: 0.02V DC.
Current Measurement adapter and probe measures current in a DC circuit. When measuring voltage at the same time, the negative terminal of each probe must be connected to the same point in the circuit.
Sensor adapter is a compact bi-polar voltage adapter.