Learn More
Brady™ Lockout Tagout Tags: DO NOT OPEN This lock/tag may only be removed by REV A
Write-On Paper Cardstock: Surface printed Cardstock (B-853) tags provide an economical, yet durable material option for temporary tagging in a non harsh environment.
Supplier: Brady™ 133558
View more versions of this product
Safety Services Available
Overview
LOTO Assessments
Description
- B-851 Rigid Economy Polyester with brass grommets, exceeds OSHA's 50 lb pull force requirement for Tagout devices
- Textured write-on surface allows for authorized signatures, dates and other special instructions
- Withstands moisture, grease and dirt
- Stays legible longer than cardstock
Specifications
Tag | |
ANSI Z53.1-1967; ANSI Z535-2006; OSHA 1910.145; OSHA 1910.147 | |
3 in. |
Economy Polyester (B-851) | |
25 Pk. | |
76.2 mm |