Learn More
Master Lock Red Anodized Aluminum Safety Padlock, 1-1/2in (38mm) Wide with 1-1/2in (38mm) Tall Shackle
Supplier: Master Lock A1106RED
This item has a minimum order quantity of 5 per supplier requirements
American Lock Aluminum Safety Padlock. 1-1/2in (38mm) wide red anodized aluminum body, 1/4in (6mm) diameter chrome plated, boron alloy shackle with 1-1/2in (38mm) clearance. Anodized finish suitable for lockout tagout in food processing facilities. Corrosion resistant finish for tough environments. High visibility color plus high security. Keyed different 5-pin tumbler rekeyable cylinder
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.