Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Eppendorf™ LoBind PCR Plates
Improve recovery of nucleic acids by reducing their absorption to the tube wall. A combination of special manufacturing technologies and selected polypropylene batches ensures nearly 100% recovery of DNA/RNA molecules-without surface coating to eliminate the risk of sample contamination.
$802.00 - $1153.00
Specifications
Format | Standard |
---|---|
Array | 16 x 24 |
Color | White |
Material | Polypropylene |
Volume (Metric) Well | 140 μL |
Catalog Number | Mfr. No. | For Use With (Application) | No. of Wells | Volume (Metric) Well | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | For Use With (Application) | No. of Wells | Volume (Metric) Well | Price | Quantity & Availability | |||
E951040589
|
Eppendorf™
951040589 |
Protein, peptide, antibody or virus samples | 384 | 140 μL |
Each for $1,153.00
|
|
|||
E951040546
|
Eppendorf™
951040546 |
DNA or RNA applications in forensic analysis, microarrays and next generation sequencing | 384 | 140 μL |
Pack of 80 for $802.00
|
|
|||
Description
- LoBind material guarantees maximum sample recovery for improved assay results
- Free of surface coating (e.g. silicone) to minimize the risk of sample interference
- Lot-certified PCR clean purity grade: of human DNA, DNAase, RNase and PCR inhibitors
- Available in tube, microplate, and deepwell plate formats for easy-up scaling
- Up to 30 % faster sample identification and fewer pipetting errors
- RecoverMax™ well design: optimized well geometry for minimal remaining/dead volume and excellent mixing properties
- Raised well rims and a smooth surface guarantee reliable sealing
Specifications
Standard | |
White | |
140 μL | |
Non-sterile | |
Yes | |
12.77 cm | |
8.55 cm |
16 x 24 | |
Polypropylene | |
384 | |
Conical Bottom (V-well) | |
Yes | |
1.44 cm |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title