Learn More
Metrohm USA LL UNITRODE PT1000
Supplier: Metrohm USA 60258600
Unitrode with Pt 1000 (without cable - electrode head U) Bridge electrolyte type: c(KCl) = 3 mol/L Diaphragm: Fixed ground-joint diaphragm Electrode plug-in head: Metrohm plug-in head U Electrode slope: > 0.97 Electrode zero point: (mV) 0 ± 15 mV (pH = 7) Electrolyte flow rate: (μl/h) 3...30 Indicator electrode shape: Cylinder Indicator electrode type: Glass electrode Internal reference electrode type: LL system Max. installation length: (mm) 113 Measuring range: 0...14 Measuring unit: pH Membrane glass type: U Membrane resistance: 150...500 Min. immersion depth: (mm) 25 Plug for filling opening: Orifice Reference electrolyte type: c(KCl) = 3 mol/L Reference system: LL system Resistance: (kOhm) 5 SGJ sleeve: flexible SGJ sleeve Shaft diameter bottom (mm): 12 Shaft diameter top (mm): 12 Shaft material: Glass Temperature range, long-term: (°C) 0 ... 80 Temperature range, short-term: (°C) 0 ... 100 Temperature sensor: Pt1000
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.