Learn More
International Crystal LITTLE PRESS PELLET DYE KIT
Supplier: International Crystal 0012193
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Little-Press Kit, Including press body, Two Type 316 Stainless Steel bolt style anvils and sample holder, The press body is a type 304 Stainless Steel nut and the anvils are Type 316 Stainless Steel bolts with optically polished ends