Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Little Joe Torso
SCI_ED

Supplier:  Anatomical Chart Company CMT4

Catalog No. SCMT4


Add to Cart

Includes: key card with 52 structures labeled.

Description

Description

A trophy-size male torso model shows musculature on one side and surface skin on the other. Dissectible into seven parts: torso, lung (two parts), heart, liver, stomach, intestine. Mounted on a wooden base. Size: 10-1/2 in. tall.

Specifications

Specifications

Model, Torso, Little Joe
26.67 cm
Wooden Base
4 lb.
10.5 in.
Dissectible parts include torso, lung (2-parts), heart, liver, stomach, intestine, key card with 52 structures labeled
Trophy-size
1.81 kg
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit