Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Lithium
SCI_ED

Click to view available options
Quantity:
25 g
Packaging:
Solid-Metal Rods
7439-93-2
Li
6.94
WHXSMMKQMYFTQS-UHFFFAOYSA-N
litio, litium, lithium, metallic, lithium, elemental, monohydride, unii-9fn79x2m3f, hsdb 647, hydride lih, hydrure de french, hsdb 549
3028194
CHEBI:30145
lithium
[Li]
7439-93-2
Li
6.94
WHXSMMKQMYFTQS-UHFFFAOYSA-N
litio, litium, lithium, metallic, lithium, elemental, monohydride, unii-9fn79x2m3f, hsdb 647, hydride lih, hydrure de french, hsdb 549
3028194
CHEBI:30145
lithium
[Li]

Reacts with water or moisture

This product is an Educational Grade Chemical — no C of A is available.

Melting Point 181°C
Boiling Point 1317°C
Density 0.534g/mL
Linear Formula Li
Quantity 25 g
Formula Weight 6.94
Packaging Solid-Metal Rods
Chemical Name or Material Lithium

ShelfLife : 5 years