Learn More
Electron Microscopy Sciences Thermo-Flask Liquid Nitrogen Transfer Vessel, 2 L, 4.25 I.D., 5.625 O.D., 10.5 height
Supplier: Electron Microscopy Sciences 6189102
Thermo-Flask provides safe extended temperature retention for specimens stored in liquid nitrogen or solid CO2, and thus have a broad range of applications in industrial, educational, and clinical fields. Thermo-Flask features borosilicate glass inner liners which have been evacuate to 1x10-5 mm of Hg to provide maximum coolant retention. Corrosion-resistant stainless steel exterior. Carrying handle, vented lid secured by clamps.
Capacity: 2 L
I.D.: 4.25
O.D.: 5.625
Height: 10.5
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.