Learn More
Alkali Scientific Liquid Nitrogen Storage System
Supplier: Alkali Scientific LN5
Liquid Nitrogen Storage System, 60 Liter capacity with stainless racks, holds 2000 x 2.0ml cryovials /EACH - fits 4 stainless steel racks, each rack 5 compartments tall with outer rod for removal - shown with optional roller base