Learn More
DIASYS DIAGOSTIC SYSTEMS USA LLC DiaSys TruLab Lipid Level 1 is a lyophilized normal lipid multi-control (3x3mL)
Supplier:  DIASYS DIAGOSTIC SYSTEMS USA LLC 590209911065
Encompass_Preferred
DiaSys TruLab Lipid Level 1 is a lyophilized normal control based on human blood material (serum) with additives of purified material of human origin. For veterinary or research use only. Contains Apolipoprotein A1, Apolipoprotein B, Cholesterol, HDL, LDL, NEFA, Phospholipids, Triglycerides, Lp-PLA2. DiaSys TruLab Lipid Level 1 must be stored refrigerated. Once reconstituted, DiaSys TruLab Lipid Level 1 is stable for 30 days when kept frozen.
Catalog No. NC2874332
Explore available promotions