Learn More
DIASYS DIAGOSTIC SYSTEMS USA LLC DiaSys TruCal Lipid is a lyophilized lipid multi-calibrator (3x2mL)
Supplier:  DIASYS DIAGOSTIC SYSTEMS USA LLC 135709911045
Encompass_Preferred
DiaSys TruCal Lipid is a lyophilized calibrator based on human blood material (plasma) with additives of purified material of human. For veterinary or research use only. Contains HDL-C, LDL-C, Lp-PLA2, NEFA and Phospholipids. DiaSys TruCal Lipid must be stored refrigerated. Once reconstituted, DiaSys TruCal Lipid is stable for 30 days when kept frozen.
Catalog No. NC2874337
Explore available promotions