Learn More
Beckman Coulter Beckman Coulter LINER KIT, HARVESTLINE, QTY 1
Supplier: Beckman Coulter 369264
Beckman Coulter HarvestLine system liner kit comes with: one bottle rack, one funnel and 120 disposable non-sterile 1L liners. (Qty 1)