Learn More
Cole-Parmer Lightnin 869580 High-Shear Dispersing Impeller, 5/16" Bore; 3.0" Dia
Supplier: Cole-Parmer 0456162
High-shear dispersing impellers create a cutting, tearing action to efficiently shred and disperse solids. Provide low flow with highest shear.