Learn More
Medicus Health Lighted Tissue Floating Bath
Supplier: Medicus Health 2627M1
∙ Lighted Tissue Floating Bath - New Lower Profile Design with Easy Grip Removable Rectangular Glass Basin, Illuminated with LED Light from the Side ∙LED Lighted rectangular basin∙Easy to read LED display shows both set temperature and actual temperature∙Temperature settings are controlled by push buttons for accuracy and precision∙CE certified under LVD 2006/95/EC∙Sleeker glass basin for better contrast of specimens. ∙Approx. ½ Gallon capacity∙Overall Size: 14"""" x 14"""" x 4.5""""; Interior Size: 10"""" x 6.5"""" x 2""""∙Temperature Range: room temp to 75C +/- 1C∙Shipping Weight: 22 lbs.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.