Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Carson Optical™ Lighted MagniGrip 4.5x Power LED Lighted Magnifier with Precision Tweezers
The Lighted MagniGrip™ is a compact 4.5x power 1.1 in. LED Lighted Magnifier with precision tweezers. This magnifier/tweezer combo is perfect for all those little jobs. It is extremely lightweight and can be used as an aid for everything from hobbies and crafts to getting out that pesky splinter.
Supplier: Carson Optical™ MG88
This item is not returnable.
View return policy
Description
The MG-88, Lighted MagniGrip™ is a compact 4.5x Power 1.1 in. LED-lighted magnifier with precision tweezers.
- This magnifier/tweezer combo is perfect for all those “little” jobs…when you need an extra hand!
- Extremely lightweight and can be used as an aid for everything from hobbies and crafts to getting out that pesky splinter. Three button cell batteries included.
Specifications
2.79 cm | |
Acrylic Lens | |
4.5x |
Magnifier | |
1.1 in. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction