Learn More
Cole-Parmer Fiber Optic Illuminator, 115 VAC
Supplier: Cole-Parmer 4150050
Illuminator only* Easy to use - simply turn it on and adjust the brightness. This low-noise illuminator features a stepless control knob to dial in the desired intensity.