Learn More
LIGATRAP TECHNOLOGIES LT Human IgG Microspin Columns- Qty 10
SDP
Supplier:  LIGATRAP TECHNOLOGIES LT095MSC
Encompass_Preferred
LigaTrap Human IgG Purification microspin columns. Includes set of ten microspin columns packed with 0.100 mL pf resin for small volume (microliters) and follow a "batch" purification method. This product is based on proprietary LigaTrap technology to utilize novel, patented ligands, designed for optimized antibody purification.
Catalog No. 50-201-6022