Learn More
Chemglass Life Sciences 7-Neck PTFE Reaction Vessel Lid Only, 80mm, Without Joint Adapters
SDP
Supplier:  Chemglass Life Sciences CG1945T11
Encompass_Preferred
7-Neck PTFE Reaction Vessel Lid Only, 80mm, Without Joint Adapters. Reaction vessel lid is constructed entirely of PTFE. Low-form lid has seven openings. Supplied WITHOUT adapters. For the complete lid, see CG-1945-T-50.
Catalog No. 50-153-1282
Explore available promotions