Learn More
Electron Microscopy Sciences LIBRA Pump, Charger, 240 V
Supplier: Electron Microscopy Sciences 6524024
The Buck LIBRA is a low cost, high performance, constant flow personal sampling pump. Suitable for use in asbestos abatement, lead sampling and industrial hygiene sampling. Designed for use with asbestos and lead abatement, the LIBRA provides full constant flow compensation using 25 mm and 37 mm cassettes. Flow rate of 0.8-4 LPM. Comes with a rechargeable NiCAD battery pack system and built-in washable stainless steel filter. It is compact, lightweight, and quiet. Full one year warranty.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.